Assay Details
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100411.2 | XP_014955897.1 | 3 - 4 | 348 | 89 | |
XM_027977743.1 | XP_027833544.1 | 3 - 4 | 289 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
