Assay Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009544.4 | XP_004009593.2 | 4 - 5 | 712 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
