Assay Details
Gene Symbol: PRAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962715.1 | XP_027818516.1 | 4 - 5 | 2507 | 110 | |
XM_027962716.1 | XP_027818517.1 | 4 - 5 | 2691 | 110 | ||
XM_027962717.1 | XP_027818518.1 | 4 - 5 | 2993 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
