Assay Details
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012165888.3 | XP_012021278.1 | 6 - 7 | 926 | 74 | |
XM_012165894.3 | XP_012021284.1 | 6 - 7 | 892 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
