Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003967.3 | XP_004004016.2 | 5 - 6 | 2160 | 61 | |
XM_015092950.2 | XP_014948436.2 | 4 - 5 | 1764 | 61 | ||
XM_015092951.2 | XP_014948437.2 | 4 - 5 | 1764 | 61 | ||
XM_027964054.1 | XP_027819855.1 | 6 - 7 | 1544 | 61 | ||
XM_027964055.1 | XP_027819856.1 | 6 - 7 | 1162 | 61 | ||
XM_027964056.1 | XP_027819857.1 | 6 - 7 | 971 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
