Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009036.3 | XP_004009085.2 | 2 - 3 | 179 | 70 | |
XM_012178886.3 | XP_012034276.2 | 2 - 3 | 154 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
