Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970662.1 | XP_027826463.1 | 3 - 4 | 349 | 96 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970662.1 | XP_027826463.1 | 3 - 4 | 349 | 96 |