Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177923.3 | XP_012033313.2 | 3 - 4 | 382 | 106 | |
XM_027970662.1 | XP_027826463.1 | 4 - 5 | 457 | 106 |

More Information
Set Membership: |
![]() ![]() |
