Assay Details
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969696.1 | XP_027825497.1 | 3 - 4 | 573 | 93 | |
XM_027969697.1 | XP_027825498.1 | 3 - 4 | 573 | 93 | ||
XM_027969698.1 | XP_027825499.1 | 3 - 4 | 573 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
