Assay Details
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969696.1 | XP_027825497.1 | 6 - 7 | 1733 | 92 | |
XM_027969697.1 | XP_027825498.1 | 7 - 8 | 1592 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
