Assay Details
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963301.1 | XP_027819102.1 | 2 - 3 | 322 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
