Assay Details
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006941.4 | XP_004006990.1 | 4 - 5 | 980 | 66 | |
XM_012175528.3 | XP_012030918.1 | 4 - 5 | 980 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
