Assay Details
Gene Symbol: LOC101117028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091984.2 | XP_014947470.2 | 1 - 2 | 306 | 85 | |
XM_027970323.1 | XP_027826124.1 | 1 - 2 | 819 | 85 | ||
XM_027970329.1 | XP_027826130.1 | 1 - 2 | 306 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
