Assay Details
Gene Symbol: RIOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002858.3 | XP_004002907.2 | 3 - 4 | 657 | 82 | |
XM_012188318.2 | XP_012043708.2 | 3 - 4 | 675 | 82 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RIOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002858.3 | XP_004002907.2 | 3 - 4 | 657 | 82 | |
XM_012188318.2 | XP_012043708.2 | 3 - 4 | 675 | 82 |