Assay Details
Gene Symbol: LOC101106395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004155.4 | XP_004004204.1 | 10 - 11 | 1823 | 61 | |
XM_027964301.1 | XP_027820102.1 | 11 - 12 | 1949 | 61 | ||
XM_027964302.1 | XP_027820103.1 | 10 - 11 | 1872 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
