Assay Details
Gene Symbol: LOC101106395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004155.4 | XP_004004204.1 | 9 - 10 | 1667 | 67 | |
XM_027964301.1 | XP_027820102.1 | 10 - 11 | 1793 | 67 | ||
XM_027964302.1 | XP_027820103.1 | 9 - 10 | 1716 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
