Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099687.2 | XP_014955173.2 | 4 - 5 | 4574 | 87 | |
XM_015099688.2 | XP_014955174.2 | 4 - 5 | 4573 | 87 | ||
XM_015099691.2 | XP_014955177.2 | 4 - 5 | 4573 | 87 | ||
XM_027976780.1 | XP_027832581.1 | 4 - 5 | 4574 | 87 | ||
XM_027976781.1 | XP_027832582.1 | 5 - 6 | 1004 | 87 | ||
XM_027976783.1 | XP_027832584.1 | 4 - 5 | 4573 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
