Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 12 - 13 | 1874 | 101 | |
XM_027975770.1 | XP_027831571.1 | 12 - 13 | 1876 | 101 | ||
XM_027975771.1 | XP_027831572.1 | 12 - 13 | 1875 | 101 | ||
XM_027975772.1 | XP_027831573.1 | 11 - 12 | 1677 | 101 | ||
XM_027975773.1 | XP_027831574.1 | 3 - 4 | 324 | 101 | ||
XM_027975774.1 | XP_027831575.1 | 3 - 4 | 203 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
