Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 14 - 15 | 2141 | 115 | |
XM_027975770.1 | XP_027831571.1 | 14 - 15 | 2143 | 115 | ||
XM_027975771.1 | XP_027831572.1 | 14 - 15 | 2112 | 115 | ||
XM_027975772.1 | XP_027831573.1 | 13 - 14 | 1944 | 115 | ||
XM_027975773.1 | XP_027831574.1 | 5 - 6 | 591 | 115 | ||
XM_027975774.1 | XP_027831575.1 | 5 - 6 | 470 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
