Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 18 - 19 | 2652 | 76 | |
XM_027975771.1 | XP_027831572.1 | 18 - 19 | 2623 | 76 | ||
XM_027975772.1 | XP_027831573.1 | 17 - 18 | 2455 | 76 | ||
XM_027975773.1 | XP_027831574.1 | 9 - 10 | 1102 | 76 | ||
XM_027975774.1 | XP_027831575.1 | 9 - 10 | 981 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
