Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 23 - 24 | 3222 | 62 | |
XM_027975770.1 | XP_027831571.1 | 23 - 24 | 3221 | 62 | ||
XM_027975771.1 | XP_027831572.1 | 23 - 24 | 3193 | 62 | ||
XM_027975772.1 | XP_027831573.1 | 22 - 23 | 3025 | 62 | ||
XM_027975773.1 | XP_027831574.1 | 14 - 15 | 1672 | 62 | ||
XM_027975774.1 | XP_027831575.1 | 14 - 15 | 1551 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
