Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 24 - 25 | 3340 | 80 | |
XM_027975770.1 | XP_027831571.1 | 24 - 25 | 3339 | 80 | ||
XM_027975771.1 | XP_027831572.1 | 24 - 25 | 3311 | 80 | ||
XM_027975772.1 | XP_027831573.1 | 23 - 24 | 3143 | 80 | ||
XM_027975773.1 | XP_027831574.1 | 15 - 16 | 1790 | 80 | ||
XM_027975774.1 | XP_027831575.1 | 15 - 16 | 1669 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
