Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 26 - 27 | 3566 | 98 | |
XM_027975770.1 | XP_027831571.1 | 26 - 27 | 3565 | 98 | ||
XM_027975771.1 | XP_027831572.1 | 26 - 27 | 3537 | 98 | ||
XM_027975772.1 | XP_027831573.1 | 25 - 26 | 3369 | 98 | ||
XM_027975773.1 | XP_027831574.1 | 17 - 18 | 2016 | 98 | ||
XM_027975774.1 | XP_027831575.1 | 17 - 18 | 1895 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
