Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 27 - 28 | 3715 | 86 | |
XM_027975770.1 | XP_027831571.1 | 27 - 28 | 3714 | 86 | ||
XM_027975771.1 | XP_027831572.1 | 27 - 28 | 3686 | 86 | ||
XM_027975772.1 | XP_027831573.1 | 26 - 27 | 3518 | 86 | ||
XM_027975773.1 | XP_027831574.1 | 18 - 19 | 2165 | 86 | ||
XM_027975774.1 | XP_027831575.1 | 18 - 19 | 2044 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
