Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 8 - 9 | 1300 | 76 | |
XM_027975770.1 | XP_027831571.1 | 8 - 9 | 1302 | 76 | ||
XM_027975771.1 | XP_027831572.1 | 8 - 9 | 1301 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
