Assay Details
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004148.4 | XP_004004197.1 | 7 - 8 | 996 | 93 |
Gene Symbol: LOC114108763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964314.1 | XP_027820115.1 | 7 - 8 | 952 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
