Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006589.3 | XP_004006638.1 | 5 - 6 | 500 | 70 | |
XM_027967604.1 | XP_027823405.1 | 4 - 5 | 442 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
