Assay Details
Gene Symbol: LOC101103343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015280.4 | XP_004015329.1 | 4 - 5 | 1122 | 70 | |
XM_004015281.4 | XP_004015330.1 | 4 - 5 | 1182 | 70 | ||
XM_027978376.1 | XP_027834177.1 | 4 - 5 | 1201 | 70 | ||
XM_027978377.1 | XP_027834178.1 | 4 - 5 | 1011 | 70 | ||
XM_027978378.1 | XP_027834179.1 | 4 - 5 | 1195 | 70 | ||
XM_027978379.1 | XP_027834180.1 | 4 - 5 | 1195 | 70 | ||
XM_027978381.1 | XP_027834182.1 | 4 - 5 | 1185 | 70 | ||
XM_027978382.1 | XP_027834183.1 | 4 - 5 | 1185 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
