Assay Details
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106715.3 | XP_011962105.2 | 2 - 3 | 862 | 71 | |
XM_027962981.1 | XP_027818782.1 | 1 - 2 | 4173 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
