Assay Details
Gene Symbol: LOC101106975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957845.1 | XP_027813646.1 | 2 - 3 | 180 | 77 | |
XM_027957846.1 | XP_027813647.1 | 4 - 5 | 295 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
