Assay Details
Gene Symbol: CST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188581.3 | XP_012043971.2 | 3 - 4 | 366 | 90 | |
XM_027976776.1 | XP_027832577.1 | 3 - 4 | 2993 | 90 |
Gene Symbol: LOC114112620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963833.1 | XP_027819634.1 | 2 - 3 | 372 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
