Assay Details
Gene Symbol: LOC114109671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958978.1 | XP_027814779.1 | 1 - 2 | 273 | 105 |
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102638.2 | XP_014958124.1 | 1 - 2 | 293 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
