Assay Details
Gene Symbol: BNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971776.1 | XP_027827577.1 | 7 - 8 | 947 | 84 | |
XM_027971777.1 | XP_027827578.1 | 8 - 9 | 1027 | 84 | ||
XM_027971778.1 | XP_027827579.1 | 7 - 8 | 1512 | 84 | ||
XM_027971779.1 | XP_027827580.1 | 7 - 8 | 947 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
