Assay Details
Gene Symbol: LOC101105610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001943.4 | XP_004001992.2 | 1 - 2 | 197 | 97 | |
XM_027964515.1 | XP_027820316.1 | 1 - 2 | 197 | 97 | ||
XM_027964517.1 | XP_027820318.1 | 1 - 2 | 197 | 97 | ||
XM_027964521.1 | XP_027820322.1 | 1 - 2 | 197 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
