Assay Details
Gene Symbol: LOC101105610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001943.4 | XP_004001992.2 | 2 - 3 | 336 | 118 | |
XM_027964511.1 | XP_027820312.1 | 2 - 3 | 324 | 118 | ||
XM_027964515.1 | XP_027820316.1 | 2 - 3 | 336 | 118 | ||
XM_027964517.1 | XP_027820318.1 | 2 - 3 | 336 | 118 | ||
XM_027964521.1 | XP_027820322.1 | 2 - 3 | 336 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
