Assay Details
Gene Symbol: LOC101105610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001943.4 | XP_004001992.2 | 9 - 10 | 1225 | 102 | |
XM_027964511.1 | XP_027820312.1 | 9 - 10 | 1213 | 102 | ||
XM_027964515.1 | XP_027820316.1 | 9 - 10 | 1129 | 102 | ||
XM_027964517.1 | XP_027820318.1 | 7 - 8 | 901 | 102 | ||
XM_027964521.1 | XP_027820322.1 | 7 - 8 | 901 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
