Assay Details
Gene Symbol: DDO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011207.4 | XP_004011256.3 | 3 - 4 | 373 | 115 | |
XM_027972371.1 | XP_027828172.1 | 3 - 4 | 546 | 115 | ||
XM_027972372.1 | XP_027828173.1 | 5 - 6 | 700 | 115 | ||
XM_027972373.1 | XP_027828174.1 | 4 - 5 | 496 | 115 | ||
XM_027972374.1 | XP_027828175.1 | 4 - 5 | 576 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
