Assay Details
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 12 - 13 | 1672 | 110 | |
XM_012101169.3 | XP_011956559.1 | 11 - 12 | 1634 | 110 | ||
XM_027958396.1 | XP_027814197.1 | 10 - 11 | 1732 | 110 | ||
XM_027958397.1 | XP_027814198.1 | 12 - 13 | 1674 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
