Assay Details
Gene Symbol: LOC114109610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958637.1 | XP_027814438.1 | 1 - 2 | 102 | 100 |
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 13 - 14 | 1842 | 100 | |
XM_012101169.3 | XP_011956559.1 | 12 - 13 | 1804 | 100 | ||
XM_027958396.1 | XP_027814197.1 | 11 - 12 | 1902 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027958637.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
