Assay Details
Gene Symbol: LOC114109610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958637.1 | XP_027814438.1 | 4 - 5 | 444 | 120 |
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 16 - 17 | 2184 | 120 | |
XM_012101169.3 | XP_011956559.1 | 15 - 16 | 2146 | 120 | ||
XM_027958396.1 | XP_027814197.1 | 14 - 15 | 2244 | 120 | ||
XM_027958397.1 | XP_027814198.1 | 14 - 15 | 1953 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
