Assay Details
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 6 - 7 | 833 | 118 | |
XM_012101169.3 | XP_011956559.1 | 5 - 6 | 795 | 118 | ||
XM_027958396.1 | XP_027814197.1 | 4 - 5 | 893 | 118 | ||
XM_027958397.1 | XP_027814198.1 | 6 - 7 | 835 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
