Assay Details
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 10 - 11 | 1396 | 71 | |
XM_012101169.3 | XP_011956559.1 | 9 - 10 | 1358 | 71 | ||
XM_027958396.1 | XP_027814197.1 | 8 - 9 | 1456 | 71 | ||
XM_027958397.1 | XP_027814198.1 | 10 - 11 | 1398 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
