Assay Details
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956609.1 | XP_027812410.1 | 2 - 3 | 154 | 74 | |
XM_027956610.1 | XP_027812411.1 | 2 - 3 | 137 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
