Assay Details
Gene Symbol: CNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012041.4 | XP_004012090.3 | 5 - 6 | 666 | 107 | |
XM_012184325.3 | XP_012039715.2 | 5 - 6 | 666 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
