Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974103.1 | XP_027829904.1 | 3 - 4 | 977 | 107 | |
XM_027974104.1 | XP_027829905.1 | 3 - 4 | 483 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
