Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974103.1 | XP_027829904.1 | 6 - 7 | 1457 | 120 | |
XM_027974104.1 | XP_027829905.1 | 5 - 6 | 861 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
