Assay Details
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972937.1 | XP_027828738.1 | 2 - 3 | 304 | 86 | |
XM_027972938.1 | XP_027828739.1 | 2 - 3 | 304 | 86 | ||
XM_027972939.1 | XP_027828740.1 | 2 - 3 | 321 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
