Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007022.4 | XP_004007071.1 | 3 - 4 | 310 | 75 | |
XM_027968159.1 | XP_027823960.1 | 3 - 4 | 294 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
