Assay Details
Gene Symbol: LOC114116817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974044.1 | XP_027829845.1 | 3 - 4 | 543 | 112 |
Gene Symbol: SPHK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974124.1 | XP_027829925.1 | 4 - 5 | 787 | 112 | |
XM_027974125.1 | XP_027829926.1 | 5 - 6 | 748 | 112 | ||
XM_027974126.1 | XP_027829927.1 | 5 - 6 | 751 | 112 | ||
XM_027974127.1 | XP_027829928.1 | 5 - 6 | 783 | 112 | ||
XM_027974128.1 | XP_027829929.1 | 5 - 6 | 684 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
