Assay Details
Gene Symbol: LOC101112335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098797.2 | XP_014954283.2 | 1 | 1309 | 81 | |
XM_027974473.1 | XP_027830274.1 | 1 | 1310 | 81 | ||
XM_027974474.1 | XP_027830275.1 | 1 | 1310 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
